Learn More
Medchemexpress LLC Lithocholic acid-d4 | 83701-16-0 | 99.9% | 380.60 | C24H36D4O3 | 10 MG
SDP
Catalog No. 5000297548
Encompass_Preferred
Lithocholic acid-d4 is the deuterium-labeled isotopologue of lithocholic acid supplied as a purified powder for use as an internal standard, tracer, or analytical reference in biochemical and analytical chemistry applications. Manufacturer-provided purity, molecular formula, and storage recommendations support use in mass spectrometry and isotope-tracing workflows.
  • Deuterium-labeled isotopologue for mass spectrometry and tracer studies.
  • High purity suitable for analytical standards and reference use.
  • Supplied as a powder with defined storage conditions for stability.
  • Molecular formula C24H36D4O3 and molecular weight 380.60 aid analytical calculations.
  • Provided in small milligram quantities for analytical workflows.
Catalog No. 50-002-97548 Supplier Medchemexpress LLC Supplier No. HYB0172S10MG
Lithocholic acid-d4 is the deuterium-labeled isotopologue of lithocholic acid supplied as a purified powder for use as an internal standard, tracer, or analytical reference in biochemical and analytical chemistry applications. Manufacturer-provided purity, molecular formula, and storage recommendations support use in mass spectrometry and isotope-tracing workflows.
  • Deuterium-labeled isotopologue for mass spectrometry and tracer studies.
  • High purity suitable for analytical standards and reference use.
  • Supplied as a powder with defined storage conditions for stability.
  • Molecular formula C24H36D4O3 and molecular weight 380.60 aid analytical calculations.
  • Provided in small milligram quantities for analytical workflows.