Learn More
Apexbio Technology LLC LITHOSPERMIC-ACID-20MG
SDP
Catalog No. 5000567681
Encompass_Preferred
lithospermic acid (CAS No 28831-65-4) is a polyphenolic compound commonly researched in the field of biomedical science for its potential therapeutic properties Originating from plants like Salvia miltiorrhiza it is known for its antioxidant and anti-inflammatory activities The mechanism of action involves the modulation of oxidative stress pathways and inhibition of inflammatory mediators making it a candidate for targeting cardiovascular diseases and metabolic disorders In vitro lithospermic acid demonstrates activity typically in the nanomolar to low micromolar range It is often utilized in various cell models and animal studies to assess its efficacy and safety profile serving as a valuable component in drug screening processes Experimental concentrations typically depend on the specific research design allowing for flexibility in application across different study types
Catalog No. 50-005-67681 Supplier Apexbio Technology LLC Supplier No. N203820
Add to Cart
Add to Cart
lithospermic acid (CAS No 28831-65-4) is a polyphenolic compound commonly researched in the field of biomedical science for its potential therapeutic properties Originating from plants like Salvia miltiorrhiza it is known for its antioxidant and anti-inflammatory activities The mechanism of action involves the modulation of oxidative stress pathways and inhibition of inflammatory mediators making it a candidate for targeting cardiovascular diseases and metabolic disorders In vitro lithospermic acid demonstrates activity typically in the nanomolar to low micromolar range It is often utilized in various cell models and animal studies to assess its efficacy and safety profile serving as a valuable component in drug screening processes Experimental concentrations typically depend on the specific research design allowing for flexibility in application across different study types