Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lithospermic acid | 28831-65-4 | 99.4% | 538.46 g·mol⁻¹ | C27H22O12 | 10 MG
SDP

Catalog No. 5000295467
Encompass_Preferred

Lithospermic acid is a plant-derived polycyclic phenolic carboxylic acid isolated from Salvia miltiorrhiza, supplied as a research reagent for biochemical and pharmacological studies including antioxidant, hepatoprotective, and enzyme inhibition investigations.

  • High purity (99.4%).
  • Solid, off-white to light yellow appearance.
  • Molecular formula C27H22O12; molecular weight 538.46 g·mol⁻¹.
  • Used in antioxidant, hepatoprotective, and enzyme inhibition research.
  • Recommended storage -20°C; protect from light; store under nitrogen.

Catalog No. 50-002-95467 Supplier Medchemexpress LLC Supplier No. HYN082310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lithospermic acid is a plant-derived polycyclic phenolic carboxylic acid isolated from Salvia miltiorrhiza, supplied as a research reagent for biochemical and pharmacological studies including antioxidant, hepatoprotective, and enzyme inhibition investigations.

  • High purity (99.4%).
  • Solid, off-white to light yellow appearance.
  • Molecular formula C27H22O12; molecular weight 538.46 g·mol⁻¹.
  • Used in antioxidant, hepatoprotective, and enzyme inhibition research.
  • Recommended storage -20°C; protect from light; store under nitrogen.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.