Learn More
Medchemexpress LLC HY-N0823 5mg Medchemexpress, Lithospermic acid CAS:28831-65-4 Purity:>98%
SDP
Catalog No. 501970312
Encompass_Preferred
Medchemexpress, HY-N0823 5mg Lithospermic acid CAS:28831-65-4 Lithospermic acid ((+)-Lithospermic acid) is a plant-derived polycyclic phenolic carboxylic acid isolated from Salvia miltiorrhiza, and has the anti-oxidative and hepatoprotective activity on carbon tetrachloride (CCl4)-induced acute liver damage in vitro and in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0312 Supplier Medchemexpress LLC Supplier No. HYN08235MG
Medchemexpress, HY-N0823 5mg Lithospermic acid CAS:28831-65-4 Lithospermic acid ((+)-Lithospermic acid) is a plant-derived polycyclic phenolic carboxylic acid isolated from Salvia miltiorrhiza, and has the anti-oxidative and hepatoprotective activity on carbon tetrachloride (CCl4)-induced acute liver damage in vitro and in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.