Learn More
Cayman Chemical LIthospermosIde 50mg
SDP
Catalog No. 502910173
Encompass_Preferred
A nitrile glucoside with antitumor activity; inhibits TPA-induced activation of EBV-EA in Raji cells
Catalog No. 50-291-0173 Supplier Cayman Chemical Supplier No. 3477150 MG
A nitrile glucoside with antitumor activity; inhibits TPA-induced activation of EBV-EA in Raji cells