Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium Diisopropylamide Solution

Catalog No. 361798800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
800 mL
4 x 100 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-179-8800ML 800 mL
36-179-8100ML 100 mL
36-179-825ML 4 x 25 mL
36-179-8 4 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 36-179-8800ML Supplier Sigma Aldrich Supplier No. 361798800ML
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lithium diisopropylamide solution (LDA) is a sterically hindered non-nucleophilic strong base widely used in organic synthesis.Lithium diisopropylamide solution (LDA) can be used:As an initiator in the anionic polymerization of D,L-lactide and methyl methacrylate.To facilitate ester enolization.To convert carboxylic acids to enediolate intermediates for preparing trifluoromethyl ketones.In ortho-lithiation of arylsulfonyloxazolidinones to prepare N-substituted saccharin analogs.To catalyze ortholithiation and Fries rearrangement of aryl carbamates.As a promoter in the isomerization of allylic ethers to (Z)-propenyl ethers.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 4111-54-0
Density 0.812 g/mL at 25°C
Molecular Formula C6H14LiN
Linear Formula [(CH3)2CH]2NLi
MDL Number MFCD00064449
Quantity 800 mL
Synonym LDA
Molecular Weight (g/mol) 107.12 g/mol
Biological Activity Hearing Organs
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.