Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Litmus, Indicator (pH 5-8), for titration, Solstice

Catalog No. 60047214 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-214 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-047-214 Supplier Solstice Supplier No. 6128025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indicator (pH 5-8), for titration

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1393-92-6
MDL Number MFCD00000826

Specifications

CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Packaging Glass Bottle
Linear Formula CHCl3
Quantity 25 g
Grade Analytical Reagent
Chemical Name or Material Litmus, Indicator (pH 5-8), for titration
ShelfLife 1620 days from date of manufacture
EINECSNumber 215-739-6
Recommended Storage Room Temp
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.