Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Litmus, MP Biomedicals™

Catalog No. MP215526381 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
150 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215526381 150 g
MP215526325 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP215526381 Supplier MP Biomedicals, Inc Supplier No. 0215526381
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Litmus, Dark blue powder

  • An acid-base indicator developed from various orcinol-containing lichen including Lecanora tartarea, Rocella fuciformis and Variolaria dealbata.
  • The principal coloring matter in Litmus is Azolitmus, a substituted 7-hydroxy-2-phenoxazone derivative.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1393-92-6
MDL Number MFCD00131528
Synonym C.I. Natural Red 28, Lackmus, Tournesole, Chhadila

Specifications

Color Dark Blue
pH 5 to 8 (literature)
Quantity 150 g
Solubility Information Soluble in ethanol (3mg/mL); partly soluble water (10mg/mL-opaque dark blue solution) or alcohol.
Physical Form Powder
storageNote1 Store at room temperature (15-30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.