Learn More
Supertek™ Litmus and pH Paper Set
GREENER_CHOICESCI_ED
Catalog No. S01270 Shop All Supertek Scientific Products
Change view
Click to view available options
pH Range:
0 to 14
Catalog No. pH Range
S01270 0 to 14
1 options
Catalog No. S01270 Supplier Supertek™ Supplier No. CH12613S3
Add to Cart
Add to Cart
The Litmus and pH Paper Set allows you to perform science experiments in your own home using common household products. Litmus paper determines whether the substance is acidic or basic; pH paper determines how acidic or basic the substance is.
  • This Litmus and PH Paper Set consists of 20 Red litmus test strips, 20 Blue litmus test strips, 40 pH test strips and 1 Activity guide.
  • Ages 7 plus
Specifications
pH Range 0 to 14