Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Litmus, pure, indicator grade

Catalog No. AC190221000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC190221000 100 g Glass bottle
AC190220250 25 g Glass bottle
AC190225000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC190221000 Supplier Thermo Scientific Chemicals Supplier No. 190221000
  • $265.00
    $225.65 / Each of 1

    Save $39.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1393-92-6

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1393-92-6
MDL Number MFCD00131528

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Visual Transition Interval from pH 4.8 (red) to pH 8.3 (blue)
Merck Index 15, 5603
Color Black/Blue to Blue
Solubility Information Solubility in water: soluble. Other solubilities: partially soluble in alcohol
Quantity 100 g
Physical Form Powder or Granules
Grade Pure
Chemical Name or Material Litmus

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.