Learn More
Cayman Chemical Litorin (trifluoroacetate salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502950007
Encompass_Preferred
A peptide with diverse biological activities; an agonist of the GRPR and the NMB receptor in BALB/c 3T3 fibroblasts expressing the human receptors (EC50s = 0.44 and 0.03 nM, respectively); binds to frog BB4 in CHO-K1 cells (Ki = 1.2 nM); induces contractions of isolated guinea pig gallbladder at 10-100 ng/kg; decreases food intake in rats at 4-128 µg/kg
Catalog No. 50-295-0007 Supplier Cayman Chemical Supplier No. 3442710 MG
A peptide with diverse biological activities; an agonist of the GRPR and the NMB receptor in BALB/c 3T3 fibroblasts expressing the human receptors (EC50s = 0.44 and 0.03 nM, respectively); binds to frog BB4 in CHO-K1 cells (Ki = 1.2 nM); induces contractions of isolated guinea pig gallbladder at 10-100 ng/kg; decreases food intake in rats at 4-128 µg/kg