Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Litronesib racemate | 546111-97-1 | 99.2% | 511.70 g·mol⁻¹ | C23H37N5O4S2 | 10 MG
SDP

Catalog No. 5000285729
Encompass_Preferred

Litronesib racemate is the racemic form of litronesib, a selective, mitosis-specific allosteric inhibitor of the kinesin Eg5 motor protein. It is supplied as a high-purity research reagent with supporting documentation for preclinical studies.

  • High purity (99.2%).
  • Selective mitosis-specific Eg5 inhibitor.
  • Molecular weight 511.70 g·mol⁻¹.
  • Chemical formula C23H37N5O4S2.
  • Available in milligram pack sizes for research use.
  • Includes data sheet, COA, and SDS.
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-85729 Supplier Medchemexpress LLC Supplier No. HY14846A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Litronesib racemate is the racemic form of litronesib, a selective, mitosis-specific allosteric inhibitor of the kinesin Eg5 motor protein. It is supplied as a high-purity research reagent with supporting documentation for preclinical studies.

  • High purity (99.2%).
  • Selective mitosis-specific Eg5 inhibitor.
  • Molecular weight 511.70 g·mol⁻¹.
  • Chemical formula C23H37N5O4S2.
  • Available in milligram pack sizes for research use.
  • Includes data sheet, COA, and SDS.
  • For research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.