Learn More
MSC Little Giant Hand Truck, 20 in Wide, 8 in Rubber Wheels, Steel, Assembled
Catalog No. 5000536633
Encompass
Little Giant hand trucks are designed for industrial use. This model features a 20-inch wide steel body with 8-inch rubber wheels, and it comes assembled for immediate use, offering a reliable solution for material handling.
  • Rigidity and longer frame life
  • Vertical center strap
  • Tubular steel for added strength
  • Equipped with rubber tires
Catalog No. 50-005-36633 Supplier MSC Supplier No. 18011841
Little Giant hand trucks are designed for industrial use. This model features a 20-inch wide steel body with 8-inch rubber wheels, and it comes assembled for immediate use, offering a reliable solution for material handling.
  • Rigidity and longer frame life
  • Vertical center strap
  • Tubular steel for added strength
  • Equipped with rubber tires