Learn More
ZORO TOOLS Little Giant Pump 1/12hp, 7 In. L, 4-1/2 In. W, 5-1/2 In. H
Catalog No. NC3782910
Encompass
HP- 1/12 hp. Voltage- 115V. Full Load Amps- 3.5 A. Impeller Material- Nylon. Duty- Continuous. Max. Head- 24.5 ft. Compatible Intake Pipe Size- 3/4 in. Compatible Discharge Pipe Size- 1/2 in
Catalog No. NC3782910 Supplier ZORO TOOLS Supplier No. G1877181 MFG504203
HP- 1/12 hp. Voltage- 115V. Full Load Amps- 3.5 A. Impeller Material- Nylon. Duty- Continuous. Max. Head- 24.5 ft. Compatible Intake Pipe Size- 3/4 in. Compatible Discharge Pipe Size- 1/2 in