Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M Littmann™ Lightweight II S.E. Stethoscope

Catalog No. 22355006
Change view
Click to view available options
Color:
Blue
Burgundy
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-355-006 Burgundy
22-355-005 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-355-006 Supplier 3M Supplier No. MMM 2451
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Littmann™ Lightweight II S.E. Stethoscope is an ideal entry level instrument, with a tunable diaphragm, dual sided chestpiece, and reliable acoustic performance. Its teardrop shape makes auscultation around blood pressure cuffs easier.

  • Reliable and clear acoustic response and light weight combine to make this an ideal entry-level instrument
  • Dual-sided chestpiece provides versatility of both a tunable diaphragm and open bell
  • Teardrop-shaped chestpiece for easier positioning under blood pressure cuffs and around bandages
  • Tunable diaphragm: Hear high or low frequency sounds by slightly adjusting pressure on the chestpiece
  • Nonchill rim and diaphragm provide welcome patient comfort
  • Two-year warranty
  • Soft-sealing eartips provide an excellent acoustic seal and comfortable fit
  • The choice for nurses

Specifications

Product Type Stethoscope
Color Burgundy
For Use With (Application) Physical Assessment and Blood Pressure
Weight (Metric) 118 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.