Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Littorine hydrochloride | 21956-47-8 | >95.0% | 325.83 | C17H24ClNO3 | 5 MG
SDP

Catalog No. 5000284747
Encompass_Preferred

Littorine hydrochloride is the hydrochloride salt of littorine, an alkaloid found in Anthocercis fittorea and Datura species. It is supplied as a small-scale research chemical with documented molecular and solubility data for in vitro preparation.

  • Hydrochloride salt of littorine, an alkaloid found in plant species.
  • Molecular formula C17H24ClNO3; molecular weight 325.83.
  • Soluble in DMSO at 50 mg/mL (153.45 mM); may require ultrasonic treatment.
  • Available in small research quantities (1 mg, 5 mg, 10 mg).
  • Store under conditions recommended in the certificate of analysis.

Catalog No. 50-002-84747 Supplier Medchemexpress LLC Supplier No. HYW6539545MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Littorine hydrochloride is the hydrochloride salt of littorine, an alkaloid found in Anthocercis fittorea and Datura species. It is supplied as a small-scale research chemical with documented molecular and solubility data for in vitro preparation.

  • Hydrochloride salt of littorine, an alkaloid found in plant species.
  • Molecular formula C17H24ClNO3; molecular weight 325.83.
  • Soluble in DMSO at 50 mg/mL (153.45 mM); may require ultrasonic treatment.
  • Available in small research quantities (1 mg, 5 mg, 10 mg).
  • Store under conditions recommended in the certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.