Learn More
Bioworld Litvay Medium, 25 L
Catalog No. 50254646
Encompass_Preferred
Litvay MediumContains Macronutrients and Micronutrients as described by Litvay et al. (1985) Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of medium Plant tissue culture tested
Catalog No. 50-254-646 Supplier Bioworld Supplier No. 306300483
Litvay MediumContains Macronutrients and Micronutrients as described by Litvay et al. (1985) Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of medium Plant tissue culture tested