Learn More
Bioworld Litvay Medium w/ Vitamins, 50 L
Catalog No. 50254650
Encompass_Preferred
Litvay MediumContains Macronutrients, Micronutrients, and Vitamins as described by Litvay et al. (1985) Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested
Catalog No. 50-254-650 Supplier Bioworld Supplier No. 306300494
Litvay MediumContains Macronutrients, Micronutrients, and Vitamins as described by Litvay et al. (1985) Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested