Learn More
Bioworld Litvay Vitamin Concentrate (Syringe Vials), 100 mL
Supplier:  Bioworld 306300501
Encompass_Preferred
Litvay Vitamin ConcentrateContains Vitamins as described by Litvay et al. (1985)Package includes 10 prepared, filtered, and sterilized ampules of 10 mL of 1000X vitamin stock solution Use 1 mL per liter of mediumContents: myo-Inositol 100.00 mg/L Nicotinic Acid 0.50 mg/L Pyridoxine HCl 0.50 mg/LThiamine HCl 0.10 mg/L Total: 5048.42 mg/L
Catalog No. 50-254-651