Learn More
Bioworld Litvay Vitamin Mix, 100 mL
Catalog No. 50254652
Encompass_Preferred
Litvay Vitamin MixContains Vitamins as described by Litvay et al. (1985)Package contains 10.07 g vitamins to prepare 100 mL of 1000X vitamin stock solution Use 1 mL per liter of mediumContents: myo-Inositol: 100.00 mg/LNicotinic Acid: 0.50 mg/LPyridoxine HCl: 0.50 mg/L Thiamine HCl: 0.10 mg/L Total: 5048.42 mg/L
Catalog No. 50-254-652 Supplier Bioworld Supplier No. 306300511
Litvay Vitamin MixContains Vitamins as described by Litvay et al. (1985)Package contains 10.07 g vitamins to prepare 100 mL of 1000X vitamin stock solution Use 1 mL per liter of mediumContents: myo-Inositol: 100.00 mg/LNicotinic Acid: 0.50 mg/LPyridoxine HCl: 0.50 mg/L Thiamine HCl: 0.10 mg/L Total: 5048.42 mg/L