Learn More
ABBERIOR INSTRUMENTS AMERICA LLC LIVE 610 tubulin (Cabazitaxel)
Supplier:  ABBERIOR INSTRUMENTS AMERICA LLC LV610014150UG
Encompass_Preferred
abberior LIVE 610 is a member of a new class of fluorescent dyes for live-cell imaging exclusively offered by abberior. This dye is highly photostable and bright red fluorescent dye, which can be effectively excited with excitation light between 630 - 650 nm. For STED, a depletion wavelength between 750 - 800 nm is recommended. Dedicated selection of the optimal isomeric structure leads to unrivaled cell permeability of this dye. abberior LIVE 610 can therefore be used in very low nanomolar concentration, reducing background and cytotoxicity effectively, allowing long-term STED and confocal imaging of living cells or tissue. Our abberior LIVE 610 is the ideal partner for abberior LIVE 550 or 560 to obtain optimal 2 color live-cell imaging results. For scientific use only. Must not be used for diagnostic or medical purposes. Cell permeable and highly specific fluorescent probe used to label tubulin in living cells.
Catalog No. NC1933888