Learn More
Cayman Chemical Live Cell Multiplex Assay, 960 Well
SDP
Catalog No. 502969840
Encompass_Preferred
The Live Cell Multiplex (LCM) Assay provides an efficient fluorescence-based method of quantifying the relative number of live cells resident in treated wells of an assay plate. While the LCM Assay may be performed as a stand-alone assay, it is has been specifically optimized to be run in multiplex with any of INDIGO’s 96-well, 2x48-well, or 3x32-well Nuclear Receptor Reporter Assay System products. [INDIGO Catalog Nos. LCM01, LCM05, LCM10]
Catalog No. 50-296-9840 Supplier Cayman Chemical Supplier No. 15766960 WELL
The Live Cell Multiplex (LCM) Assay provides an efficient fluorescence-based method of quantifying the relative number of live cells resident in treated wells of an assay plate. While the LCM Assay may be performed as a stand-alone assay, it is has been specifically optimized to be run in multiplex with any of INDIGO’s 96-well, 2x48-well, or 3x32-well Nuclear Receptor Reporter Assay System products. [INDIGO Catalog Nos. LCM01, LCM05, LCM10]