Learn More
Bioptechs™ Live Cell Microscopy Delta T™ Culture Dish System
Catalog No. 12071205
Click to view available options
Type:
Delta T Starter Set
TPG Culture Dish
Color:
Black
Clear
N/A
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Color
12-071-205 Delta T Starter Set N/A
12-071-34 TPG Culture Dish Black
12-071-33 TPG Culture Dish Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-071-205 Supplier Bioptechs™ Supplier No. 04204
Add to Cart
Add to Cart
Includes
  • Internal reference for user-adjustable calibration and TTL interface for confocal application.
  • Delta T Starter Set includes: Delta T Controller; 10 Pk. of Delta T Dishes, 0.17mm thick; Coverglass Lid; Heated Lid with CO2 port; Culture Cylinder Pack, 4mm, 6mm, 8mm, 10mm; Standard Stage Adapter (specify when ordering).
Component of Bioptechs open and closed systems for live-cell micro-observations and time-lapse imaging. Open-dish, live-cell environmental/optical containment system
For micro-observation and applications requiring physical access to cells or tissue: microinjection, electrostimulation, wounding, micromanipulation, mechanical probing, recording, and stimulation
  • Accurate temperature control, uniform heat distribution, and high-numeric-aperture compatibility
  • Works in confocal applications; adapts to specimens from monolayered adherent cells to tissue preparations, with inverted, water-immersion, and upright microscopes
  • System passes ∽0.9w electrical charge through thin indium tin-oxide coating on underside of glass substrate where cells are grown, transferring heat through glass to cells
  • Thermal regulation via instantaneous analog control of a feedback loop
  • No peripheral heating: cells are unaffected by surface evaporation, heat is not lost to the atmosphere
  • After plating, cells are ready for incubation or time-lapse imaging
  • Controller has real-time temperature display (ambient to 50°C, accuracy ±0.1°C); compensates for surface evaporation
  • Alarmed protection circuit safeguards cells. Dimensions: 151/2L x 6W x 3 in. H (39.4 x 15.2 x 7.6cm). Operates on 100–240V 50/60Hz or 24V battery
Delta T Culture Dish options
  • Disposable polystyrene/glass dishes are available plain (when temperature control is not needed) or with electrically conductive, optically transparent coating and silver bus bars; 6mm depth optimizes access for microinjection and manipulation
  • Contain self heating ITO coating, which provides first surface heat transfer directly to cells when using Delta T System
  • Injection molded polystyrene sidewalls and lid, coverglass bottom
  • Highest quality German Desag 263, low alkaline coverglass for optimum resolution
  • O.D., 35mm, tapering to 23mm optical aperture
  • Glass thickness of 0.17mm
  • Available in clear and black; for fluorescence imaging, select black culture dishes
  • Numerous accessories specific to these dishes allow for specialized applications including - media perfusion, gas perfusion, sample positioning, heating and cooling
  • Optional stage adapters available to fit all major brands of microscopes, which custom stage adapters available
  • Optional culturing cylinders may be used to barricade cells or suspended specimens in a dish; or to restrict and concentrate growth and location of cells plated on coverslip without use of grease
Specifications
Type Delta T Starter Set
Color N/A
For Use With (Application) Micro observation and applications requiring physical access to cells or tissue, microinjection, electrostimulation, wounding, micromanipulation, mechanical probing, recording and stimulation
Includes Delta T controller, delta T dishes, 0.17mm thick coverglass lid, heated lid with CO2 port, culture cylinder Pack, 4mm, 6mm, 8mm, 10mm, standard stage adapter