Learn More
Bioptechs™ Live Cell Microscopy Perfusion System
Catalog No. 1207137
Encompass
Change view
Click to view available options
Type:
Perfusion Connector Kit
Catalog No. Type
12-071-37 Perfusion Connector Kit
1 options
Catalog No. 12-071-37 Supplier Bioptechs™ Supplier No. 1620031
Includes
  • 9V AC adapter; contains an internal 9V battery that can function as primary power
  • Single 1/16 in. I.D. tube for use as a single-channel pump and a dual tube with two 1/16 in. I.D. tubes for use as a dual-channel pump
  • Tubes are terminated with a 1/16 in. tubing barb
Component of Bioptechs open and closed systems for live-cell micro-observations and time-lapse imaging. Required for use with Bioptechs FCS2 closed system
  • Delta T open-dish system requires hinged perfusion adapter set, perfusable cover glass lid, microperfusion pump, and double tube pump set
  • FCS2 and FCS3 closed systems require microperfusion pump and single tube pump set
Microperfusion Pump
  • Single- or dual-channel peristaltic pump designed specifically for low flowrates of 2 to 75mL/hr.
  • Pump base is threaded for easy mounting to a stand or fixture near the microscope
  • Driven by a tachometer-regulated, multistage DC gear motor for smooth analog rotation of the roller spindle to prevent instantaneous steps
  • Regulated by internal control circuitry (adjustable from 2 to 75mL/hr) or by computer via computer-controlled analog port
Order Info
Pump measures 4L x 2 1/2W x 2 1/4 in.H (10 x 6 x 6cm); weighs 1 lb. (0.45kg).
Specifications
Type Perfusion Connector Kit
Includes 9V AC adapter, contains an internal 9V battery that can function as primary power, single 1/16 in. I.D. tube for use as a single-channel pump and a dual tube with two 1/16 in. I.D. tubes for use as a dual-channel pump, tubes are terminated with a 1/16 in. tubing barb
Voltage 9 V