Learn More
Indigo Biosciences Live Cell Multiplex (LCM) Cytotoxicity Assay Kit, 10x96-well format assays
Catalog No. 502231848
Encompass_Preferred
The Live Cell Multiplex (LCM) Assay provides an efficient fluorescence-based method of quantifying the relative number of live cells resident in treated wells of an assay plate. While the LCM Assay may be performed as a stand-alone assay, it has been specifically optimized to be run in multiplex with any of INDIGO’s 96-well, 2x48-well, or 3x32-well Reporter Assay System products.The LCM assay allows users to validate their primary assay data by determining if their test compound treatments exert mitogenic, cytostatic or cytotoxic activities on reporter cells. The occurrence of such adverse non-specific effects will always undermine the accurate assessment of a test compound’s potency and/or efficacy as a modulator of nuclear receptor function.
This item is not returnable. View return policy
Catalog No. 50-223-1848 Supplier Indigo Biosciences Supplier No. LCM10
The Live Cell Multiplex (LCM) Assay provides an efficient fluorescence-based method of quantifying the relative number of live cells resident in treated wells of an assay plate. While the LCM Assay may be performed as a stand-alone assay, it has been specifically optimized to be run in multiplex with any of INDIGO’s 96-well, 2x48-well, or 3x32-well Reporter Assay System products.The LCM assay allows users to validate their primary assay data by determining if their test compound treatments exert mitogenic, cytostatic or cytotoxic activities on reporter cells. The occurrence of such adverse non-specific effects will always undermine the accurate assessment of a test compound’s potency and/or efficacy as a modulator of nuclear receptor function.