Learn More
Apexbio Technology LLC Live-Dead Cell Staining Kit, 500T. Cas: MFCD: NA.
SDP
Catalog No. 501998365
Encompass_Preferred
Our product Live-Dead Cell Staining Kit is used for simultaneous fluorescence staining of viable and dead cells. This kit contains two dyes - Calcein-AM and Propidium Iodide (PI), which stain viable and dead cells, respectively.
Catalog No. 50-199-8365 Supplier Apexbio Technology LLC Supplier No. K2081500
Our product Live-Dead Cell Staining Kit is used for simultaneous fluorescence staining of viable and dead cells. This kit contains two dyes - Calcein-AM and Propidium Iodide (PI), which stain viable and dead cells, respectively.