Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Live Moina
SDPSCI_ED

Catalog No. S42843 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Sufficient For:
Class of 30 students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
S42843 Class of 30 students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S42843 Supplier Carolina™ Supplier No. 142336

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Acts as good live food for freshwater larval-stage fish.

  • Belongs to Daphnid family but much smaller than Daphnia pulex (approximately half the maximum length)
  • Cultured on freshwater algae

Specifications

For Use With (Application) Live food for freshwater larval-stage fish
Packaging Jar
Quantity 1 Ea.
Scientific Name Moina
Sufficient For Class of 30 students
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.