Learn More
Aat Bioquest Live or Dead Cell Viability Assay Kit Green/Red Dual Fluorescence (200 Tests)
SDP
Catalog No. 502250124
Encompass_Preferred
This Live or Deadcell viability uses two fluorogenic indicators calcein AM for viable cells and a cell-impermeable DNA-binding dye for the cells with compromised membranes. This catalog is for 200 Tests.
This item is not returnable. View return policy
Catalog No. 50-225-0124 Supplier Aat Bioquest Supplier No. 22789
This Live or Deadcell viability uses two fluorogenic indicators calcein AM for viable cells and a cell-impermeable DNA-binding dye for the cells with compromised membranes. This catalog is for 200 Tests.