Learn More
Biotium Live-or-Dye™ Staining Kit 488/515 50 Assays
Supplier:  Biotium 32004T
Encompass_Preferred
Live-or-Dye™ Fixable Viability Staining Kits are cell membrane impermeable amine-reactive dyes designed for discrimination between live and dead cells during flow cytometry. The dyes are able to enter into dead cells that have compromised membrane integrity and covalently label free amines on intracellular proteins. The dye labeling is extremely stable, allowing the cells to be fixed and permeabilized without loss of fluorescence or dye transfer between cells. The Live-or-Dye™ staining protocol has been optimized to maximize live/dead discrimination with minimal live cell staining, in order to prevent interference with immunostaining. Live-or-Dye™ Fixable Viability Stains are dead cell specific in all cell types, including mammalian cells, bacteria and yeast. Live-or-Dye™ 488/515 is suitable for excitation by 488 nm laser line. This trial size kit has sufficient reagent for 50 assays.
Catalog No. 50-196-4595