Learn More
Biotium Live-or-Dye™ 594/614 Fixable Viability Staining Kit, Trial Size (50 assays)
Catalog No. 501964599
Encompass_Preferred
Live-or-Dye™ Fixable Viability Staining Kits are cell membrane impermeable amine-reactive dyes designed for discrimination between live and dead cells during flow cytometry. The dyes are able to enter into dead cells that have compromised membrane integrity and covalently label free amines on intracellular proteins. The dye labeling is extremely stable, allowing the cells to be fixed and permeabilized without loss of fluorescence or dye transfer between cells. The Live-or-Dye™ staining protocol has been optimized to maximize live/dead discrimination with minimal live cell staining, in order to prevent interference with immunostaining. Live-or-Dye™ Fixable Viability Stains are dead cell specific in all cell types, including mammalian cells, bacteria and yeast. Live-or-Dye™ 594/614 is suitable for excitation by 488 nm or 561 nm laser line. This trial size kit has sufficient reagent for 50 assays.
Catalog No. 50-196-4599 Supplier Biotium Supplier No. 32006T
Add to Cart
Add to Cart
Live-or-Dye™ Fixable Viability Staining Kits are cell membrane impermeable amine-reactive dyes designed for discrimination between live and dead cells during flow cytometry. The dyes are able to enter into dead cells that have compromised membrane integrity and covalently label free amines on intracellular proteins. The dye labeling is extremely stable, allowing the cells to be fixed and permeabilized without loss of fluorescence or dye transfer between cells. The Live-or-Dye™ staining protocol has been optimized to maximize live/dead discrimination with minimal live cell staining, in order to prevent interference with immunostaining. Live-or-Dye™ Fixable Viability Stains are dead cell specific in all cell types, including mammalian cells, bacteria and yeast. Live-or-Dye™ 594/614 is suitable for excitation by 488 nm or 561 nm laser line. This trial size kit has sufficient reagent for 50 assays.