Learn More
Biotium Live-or-Dye™ 640/662 Fixable Viability Staining Kit | 200 assays
Catalog No. NC1700066
Encompass_Preferred
Live-or-Dye™ Fixable Viability Staining Kits allow discrimination between live and dead cells by flow cytometry or microscopy. Live/dead stains are useful probes to include when analyzing cell surface protein expression by flow cytometry, because they allow intracellular fluorescence signal from dead cells with permeable plasma membranes to be excluded from analysis.
Catalog No. NC1700066 Supplier Biotium Supplier No. 32007
Live-or-Dye™ Fixable Viability Staining Kits allow discrimination between live and dead cells by flow cytometry or microscopy. Live/dead stains are useful probes to include when analyzing cell surface protein expression by flow cytometry, because they allow intracellular fluorescence signal from dead cells with permeable plasma membranes to be excluded from analysis.