Learn More
Bachem Liver Cell Growth Factor Acetate
SDP
Supplier:  Bachem 4000308.1000
Encompass_Preferred
Liver Cell Growth Factor Acetate, H-Gly-His-Lys-OH, 72957-37-0, C14H24N6O4, Mr 340.38, 1g. GHK, Prezatide. GHK possesses a high affinity for copper(II) ions, with which it spontaneously forms a complex (GHK-Cu). This growth-modulating plasma tripeptide, when added at nanomolar concentrations to a wide group of cultured systems, produces a disparate set of responses ranging from the stimulation of growth and differentiation to outright toxicity. It may function by facilitating copper uptake into cells. GHK-Cu accelerates wound healing. CAS Number (net): 49557-75-7.
Catalog No. 50-382-803