Learn More
CPC Scientific H-Gly-His-Lys-OH (acetate salt) 1MG
SDP
Catalog No. 502109406
Encompass
GHK possesses a high affinity for copper(II) ions, with which it spontaneously forms a complex (GHK-Cu). This growth-modulating plasma tripeptide, when added at nanomolar concentrations to a wide group of cultured systems, produces a disparate set of responses ranging from the stimulation of growth and differentiation to outright toxicity. It may function by facilitating copper uptake into cells. GHK-Cu accelerates wound healing. The palmitoylated peptide is marketed as a cosmetic ingredient under the trade name Biopeptide CL. CAS Number (net): 49557-75-7.SEQUENCE: H-Gly-His-Lys-OH (acetate salt)ONE-LETTER SEQUENCE: GHKMOLECULAR FORMULA: C14H24N6O4MOLECULAR WEIGHT: 340.38STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [72957-37-0]SYNONYMS: Prezatide, GHKRESEARCH AREA: MiscellaneousREFERENCES:S.-J.Lau and B.Sarkar, Biochem. J., 199, 649 (1981)J.-P.Laussac et al., Biochem. J., 209, 533 (1983)D.Downey et al., Surg. Forum, 36, 573 (1985)L.Pickart and S.Lovejoy, Me
Catalog No. 50-210-9406 Supplier CPC Scientific Supplier No. GRFF008A
GHK possesses a high affinity for copper(II) ions, with which it spontaneously forms a complex (GHK-Cu). This growth-modulating plasma tripeptide, when added at nanomolar concentrations to a wide group of cultured systems, produces a disparate set of responses ranging from the stimulation of growth and differentiation to outright toxicity. It may function by facilitating copper uptake into cells. GHK-Cu accelerates wound healing. The palmitoylated peptide is marketed as a cosmetic ingredient under the trade name Biopeptide CL. CAS Number (net): 49557-75-7.SEQUENCE: H-Gly-His-Lys-OH (acetate salt)ONE-LETTER SEQUENCE: GHKMOLECULAR FORMULA: C14H24N6O4MOLECULAR WEIGHT: 340.38STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [72957-37-0]SYNONYMS: Prezatide, GHKRESEARCH AREA: MiscellaneousREFERENCES:S.-J.Lau and B.Sarkar, Biochem. J., 199, 649 (1981)J.-P.Laussac et al., Biochem. J., 209, 533 (1983)D.Downey et al., Surg. Forum, 36, 573 (1985)L.Pickart and S.Lovejoy, Me