Learn More
Gibco™ Liver Digest Medium

Description
Liver Digest Medium is a collagenase-dispase medium for dissociation of viable liver cells.
Specifications
Specifications
| Cell Type | Hepatocyte Cells |
| Classification | Serum-free |
| Form | Liquid |
| Product Type | Liver Digest Medium |
| Serum Level | Serum-free |
| With Additives | Phenol Red |
| Quantity | 500 mL |
| Shipping Condition | Dry Ice |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Culture Type | Mammalian Cell Culture |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.