Learn More
Lonza Walkersville Liver Mixed NPCs Human
Catalog No. 5000496350
Encompass
Human cryopreserved liver mixed NPCs are a native mixture of non-parenchymal cells from human liver These are supplied in 1million cells/vial and can have a varying amount of the different non-parenchymal cells This product is most suitable for liver cell co-culture applications
Catalog No. 50-004-96350 Supplier Lonza Walkersville Supplier No. HUCNP
Human cryopreserved liver mixed NPCs are a native mixture of non-parenchymal cells from human liver These are supplied in 1million cells/vial and can have a varying amount of the different non-parenchymal cells This product is most suitable for liver cell co-culture applications