Learn More
Electron Microscopy Sciences LiveSlide Microscope Living Slides
SDP
Supplier:  Electron Microscopy Sciences 7187750
Encompass_Preferred
These innovative, optically clear microscope slides have a shallow reservoir that confines microorganisms to a liquid depth not much larger than the depth of field of the microscope, yet deep enough to allow the microscopic animals to move about freely. Large viewing area has partitions to keep specimens in viewing area and uniform depth of 0.009".
Made of Makrolon 2558 polycarbonate and includes two reusable coverslips made of Makrofol, 0.005" and 0.010". Both the base and the coverslips are reusable. Features excellent impact resistance for safety indoors and outdoors. It can be used as a traditional slide simply by flipping the base over. 100% made in the USA.
Catalog No. 50-193-0741