Learn More
Electron Microscopy Sciences LiveSlide Microscope Living Slides
SDP
Catalog No. 501930739
Encompass_Preferred
These innovative, optically clear microscope slides have a shallow reservoir that confines microorganisms to a liquid depth not much larger than the depth of field of the microscope, yet deep enough to allow the microscopic animals to move about freely. Large viewing area has partitions to keep specimens in viewing area and uniform depth of 0.009".
Made of Makrolon 2558 polycarbonate and includes two reusable coverslips made of Makrofol, 0.005" and 0.010". Both the base and the coverslips are reusable. Features excellent impact resistance for safety indoors and outdoors. It can be used as a traditional slide simply by flipping the base over. 100% made in the USA.
Catalog No. 50-193-0739 Supplier Electron Microscopy Sciences Supplier No. 7187705
These innovative, optically clear microscope slides have a shallow reservoir that confines microorganisms to a liquid depth not much larger than the depth of field of the microscope, yet deep enough to allow the microscopic animals to move about freely. Large viewing area has partitions to keep specimens in viewing area and uniform depth of 0.009".
Made of Makrolon 2558 polycarbonate and includes two reusable coverslips made of Makrofol, 0.005" and 0.010". Both the base and the coverslips are reusable. Features excellent impact resistance for safety indoors and outdoors. It can be used as a traditional slide simply by flipping the base over. 100% made in the USA.