Learn More
Description
- Storage: Some Components Require Refrigerator and Freezer Storage Upon Receipt
- Time Required: 60 Minutes (plus two incubations)
- For 25 Students
Specifications
Specifications
| Product Type | Kit |
| Grade | 11 to Undergraduate |
| Age | 16 to 20 |
| For Use With (Equipment) | Edvotek™ Transformation Kits 222, 223, 224 or 303 |
| Includes | Instructions, Ampicillin, IPTG, GFP BactoBeads™, ReadyPour™ Luria Broth Agar (sterile), Luria Broth Liquid Media, Inoculating Loops (sterile), Plastic Loops (non-sterile), Toothpicks, Petri Plates, 1.5 mL Microcentrifuge Tubes, 10 mL Pipette (sterile) |
| Requires | Transformed Colonies from Edvotek™ Cat. 222/223/224/303, Distilled Water, Gloves, UV Light Source, Microwave, Incubator or Water Bath, Adjustable Pipette and Tips (20-200 μL) |
| Time Range | 60 Minutes (plus two incubations) |
| Content And Storage | Some Components Require Refrigerator and Freezer Storage Upon Receipt |
| Class Size | 25 Students |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

