Learn More
Genscript Corporation LIX/CXCL5 (92aa), Mouse
Supplier:  Genscript Corporation Z028511
Encompass
The mouse homolog of ENA-78 is called LIX. ENA-78/LIX is a CXC chemokine that signals through the CXCR2 receptor. It is expressed in monocytes, platelets, endothelial cells, and mast cells. ENA-78/LIX is a chemoattractant for neutrophils. The three naturally occurring variants of human ENA-78; ENA 5-78, ENA 9-78 and ENA 10-78, contain 74, 70, and 69 amino acid residues, respectively, and possess the same biological activity. ENA-78/LIX contains the four conserved cysteine residues present in CXC chemokines, and also contains the 'ELR' motif common to CXC chemokine that bind to the CXCR1 and CXCR2 receptors.
Catalog No. 50-276-682