Learn More
Aviva Systems Biology Corp LIX1L Antibody - N-terminal region (ARP53200_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53200_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LIX1L. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human LIX1L. Alternate names: DKFZp762F237, MGC46719ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-807