Learn More
Medchemexpress LLC HY-P0119A 1mg Medchemexpress, Lixisenatide (acetate) CAS:1997361-87-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0119A1MG
Encompass_Preferred
Medchemexpress, HY-P0119A 1mg Lixisenatide (acetate) CAS:1997361-87-1 Lixisenatide acetate is a glucagon-like peptide-1 (GLP-1) receptor agonist that can be used in the treatment of type 2 diabetes mellitus (T2DM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1649