Learn More
Cayman Chemical LIxIsenatIdeacetate 50mg
SDP
Catalog No. 502941976
Encompass_Preferred
A GLP-1R agonist; binds to CHO-K1 cells expressing human GLP-1R (IC50 = 1.4 nM); inhibits Il-1β- and IFN-γ-induced apoptosis in INS-1 rat pancreatic β-cells at 100 pM; lowers blood glucose levels in an OGTT (ED50 = 0.021 nmol/kg) and decreases blood HbA1c levels in db/db mice
Catalog No. 50-294-1976 Supplier Cayman Chemical Supplier No. 3973950 MG
A GLP-1R agonist; binds to CHO-K1 cells expressing human GLP-1R (IC50 = 1.4 nM); inhibits Il-1β- and IFN-γ-induced apoptosis in INS-1 rat pancreatic β-cells at 100 pM; lowers blood glucose levels in an OGTT (ED50 = 0.021 nmol/kg) and decreases blood HbA1c levels in db/db mice