Learn More
Medchemexpress LLC HY-14185 2mg Medchemexpress, Lixivaptan CAS:168079-32-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY141852MG
Encompass_Preferred
Medchemexpress, HY-14185 2mg Lixivaptan CAS:168079-32-1 Lixivaptan (VPA-985, WAY-VPA 985) is an orally active and selective vasopressin receptor V2 antagonist, with IC50 values of 1.2 and 2.3 nM for human and rat V2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1573