Learn More
TARGETMOL CHEMICALS INC LJ-001 5MG
SDP
Catalog No. 502580322
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LJ-001 shows broad-spectrum activity against virtually all enveloped RNA and DNA viruses. LJ001 inhibits the infection of transmissible gastroenteritis virus (TGEV) and porcine delta coronavirus (PDCoV). purity: 98%
Catalog No. 50-258-0322 Supplier TARGETMOL CHEMICALS INC Supplier No. T328045MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LJ-001 shows broad-spectrum activity against virtually all enveloped RNA and DNA viruses. LJ001 inhibits the infection of transmissible gastroenteritis virus (TGEV) and porcine delta coronavirus (PDCoV). purity: 98%