Learn More
Medchemexpress LLC HY-19712 5mg Medchemexpress, LJH685 CAS:1627710-50-2 Purity:>98%
SDP
Catalog No. 502030871
Encompass_Preferred
Medchemexpress, HY-19712 5mg LJH685 CAS:1627710-50-2 LJH685 is a potent, specific and selective RSK inhibitor, inhibits RSK1, 2, and 3 biochemical activities with IC50s of 6, 5, 4 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0871 Supplier Medchemexpress LLC Supplier No. HY197125MG
Medchemexpress, HY-19712 5mg LJH685 CAS:1627710-50-2 LJH685 is a potent, specific and selective RSK inhibitor, inhibits RSK1, 2, and 3 biochemical activities with IC50s of 6, 5, 4 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.