Learn More
Selleck Chemical LLC LJH685 5mg 1627710-50-2
SDP
Catalog No. NC1147226
Encompass_Preferred
LJH685 is a potent pan-RSK inhibitor with IC50 of 6 nM, 5 nM and 4 nM for RSK1, RSK2, and RSK3, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1147226 Supplier Selleck Chemical LLC Supplier No. S78705MG
LJH685 is a potent pan-RSK inhibitor with IC50 of 6 nM, 5 nM and 4 nM for RSK1, RSK2, and RSK3, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.