Learn More
TARGETMOL CHEMICALS INC LJP-1586 HCL 5MG
SDP
Catalog No. 502579040
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. LJP-1586 HCl a highly selective inhibitor of vascular adhesion protein-1 (VAP-1) exhibits antiinflammation effect by reducing adhesion molecule expression and immune cell infiltration after intracerebral hemorrhage (ICH). purity: 99%
Catalog No. 50-257-9040 Supplier TARGETMOL CHEMICALS INC Supplier No. T88575MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. LJP-1586 HCl a highly selective inhibitor of vascular adhesion protein-1 (VAP-1) exhibits antiinflammation effect by reducing adhesion molecule expression and immune cell infiltration after intracerebral hemorrhage (ICH). purity: 99%