Learn More
EPSON LabelWorks Ribbon LK Cartridge ~3/4" Black on Skyblue
SDP
Catalog No. NC3445255
Encompass
Create extraordinary labels with Epson LabelWorks LK Tape Cartridges.Nearly 16' of tape (5 m)Great for your adding a personal touch to cards, crafts, and moreUse scissors to cut this fabric tape for the best results.Do not use the Auto-cut, Half cut or Auto-trimmer (not available for all printers).Set the low-speed printing mode if available on printer.
Catalog No. NC3445255 Supplier EPSON Supplier No. LK5LBK
Create extraordinary labels with Epson LabelWorks LK Tape Cartridges.Nearly 16' of tape (5 m)Great for your adding a personal touch to cards, crafts, and moreUse scissors to cut this fabric tape for the best results.Do not use the Auto-cut, Half cut or Auto-trimmer (not available for all printers).Set the low-speed printing mode if available on printer.