Learn More
MSC LK Flat Bag, 5 x 8 in, 2 mil, 2000/Case
Catalog No. 5000788647
Encompass
These low-density polyethylene (LDPE) flat bags provide a common packaging and storage solution for various light-duty applications. Constructed from virgin materials and seamless tubing, they resist punctures and tears, ensuring contents remain visible and protected. They meet FDA requirements for use in food applications.
  • Made from seamless tubing to prevent rips
  • Packaged in dispenser packs of 100 for convenience
  • Complies with FDA requirements for food applications
  • Constructed from clear, low-density polyethylene (LDPE)
  • Flat, open-ended design
  • Thickness of 2 mil resists punctures and tears
  • Features 5 x 8 in dimensions
Catalog No. 50-007-88647 Supplier MSC Supplier No. 39963905
These low-density polyethylene (LDPE) flat bags provide a common packaging and storage solution for various light-duty applications. Constructed from virgin materials and seamless tubing, they resist punctures and tears, ensuring contents remain visible and protected. They meet FDA requirements for use in food applications.
  • Made from seamless tubing to prevent rips
  • Packaged in dispenser packs of 100 for convenience
  • Complies with FDA requirements for food applications
  • Constructed from clear, low-density polyethylene (LDPE)
  • Flat, open-ended design
  • Thickness of 2 mil resists punctures and tears
  • Features 5 x 8 in dimensions