Learn More
Electron Microscopy Sciences LKB Chuck
SDP
Catalog No. 5029555
Encompass_Preferred
Precision machined from aluminum. These special chucks are made to accommodate small specimen blocks, which have been polymerized in Eppendorf tubes.
Catalog No. 50-295-55 Supplier Electron Microscopy Sciences Supplier No. 69991
Precision machined from aluminum. These special chucks are made to accommodate small specimen blocks, which have been polymerized in Eppendorf tubes.